ContactUsLinkComponent
Tubing cutter, Ridgid®
Tubing cutter, Ridgid®
Catalog # REST23011
Supplier:  Restek
undefined
Tubing cutter, Ridgid®
Catalog # REST23011
Supplier:  Restek
Specifications
  • Description:
    Ridgid tubing cutter for 3,1 to 6,35 mm metal tubing
  • Pk:
    1
Specifications
About this item
These compact cutters are suitable for cutting metal tubings.
  • Replaceable cutting wheel
  • Not for use with stainless steel tubing