Order Entry
Germany
ContactUsLinkComponent
 
Separating spatula
Catalog # HAMMHWH225-00
Supplier:  HAMMACHER KARL
undefined
Separating spatula
Catalog # HAMMHWH225-00
Supplier:  HAMMACHER KARL

Specifications

  • Length:
    175 mm
  • Seq:
    07
  • Blade width:
    3 mm
  • Pk:
    1

Specifications

About this item

Double ended separating spatula made from WIRONIT steel.