Orders LinkContactUsLinkComponent
Shell vials
Shell vials
Supplier:   Restek
Shell vials
Supplier:  Restek
Certificates
Shell vials
7 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Capacity
    Ø×H
    Material
    Colour
    Cap included
    Availability
    Price
  • REST24662
    Shell vials
    Shell vials
    1,00 ml
    8×40 mm
    Glass
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 36,00 €
    36,00 €
    1 * 200 ST
       36,00 €
  • REST24663
    Shell vials
    Shell vials
    1,00 ml
    8×40 mm
    Glass
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 145,00 €
    145,00 €
    1 * 1.000 ST
       145,00 €
  • REST24664
    Shell vials
    Shell vials
    1,00 ml
    8×40 mm
    Glass
    Amber
    No
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 39,00 €
    39,00 €
    1 * 200 ST
       39,00 €
  • REST24665
    Shell vials
    Shell vials
    1,00 ml
    8×40 mm
    Glass
    Amber
    No
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 188,00 €
    188,00 €
    1 * 1.000 ST
       188,00 €
  • REST24679
    Shell vials
    Shell vials
    1,00 ml
    8×40 mm
    PP
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 156,00 €
    156,00 €
    1 * 1.000 ST
       156,00 €
  • REST24680
    Shell vials, limited volume
    Shell vials, limited volume
    0,70 ml
    8×40 mm
    PP
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 48,00 €
    48,00 €
    1 * 200 ST
       48,00 €
  • REST24681
    Shell vials, limited volume
    Shell vials, limited volume
    0,70 ml
    8×40 mm
    PP
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 227,00 €
    227,00 €
    1 * 1.000 ST
       227,00 €
Starburst plugs
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Snap plugs for shell vials, silicone-lined
    8 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 393,00 €
    393,00 €
    1 * 1.000 ST
       393,00 €
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Snap plugs for shell vials, silicone-lined
    8 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 82,00 €
    82,00 €
    1 * 200 ST
       82,00 €
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Snap plugs for shell vials, conical
    8 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 25,00 €
    25,00 €
    1 * 200 ST
       25,00 €
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Snap plugs for shell vials, conical
    8 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 84,00 €
    84,00 €
    1 * 1.000 ST
       84,00 €
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Snap plugs for shell vials
    8 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 22,00 €
    22,00 €
    1 * 200 ST
       22,00 €
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Snap plugs for shell vials
    8 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 64,00 €
    64,00 €
    1 * 1.000 ST
       64,00 €