Auftragserfassung
Germany
AuftragsverfolgungKontakt
MINICYL regulators
 :   Restek
MINICYL regulators
 :  Restek
MINICYL regulators
MINICYL regulators
This compact general purpose regulator is constructed of lightweight aluminum with an elastomer diaphragm. It has many laboratory applications including air-drying glassware, sparging or evaporating solutions and controlling pneumatic valves.
Product Family Options
  • Bezeichnung
  • MINICYL regulator, 3,2 mm brass fittings, max pressure: 300 psi
    1 * 1 ST
    1 * 1 ST - 277,00 €
    277,00 €
    1 * 1 ST
       277,00 €
  • MINICYL regulator, 6,3 mm brass fittings, max pressure: 300 psi
    1 * 1 ST
    1 * 1 ST - 283,00 €
    283,00 €
    1 * 1 ST
       283,00 €