ContactUsLinkComponent
Backpressure gas regulator
Backpressure gas regulator
Catalog # REST20635
Supplier:  Restek
undefined
Backpressure gas regulator
Catalog # REST20635
Supplier:  Restek
Specifications
  • Description:
    Backpressure gas regulator
  • Pk:
    1
Specifications
About this item
Capillary GC inlet systems have backpressure regulators to maintain a constant upstream pressure and rapidly respond to catastrophic leaks. The operating range 0 to 4,1 bar is sufficient to operate a 105 m, 0,25 mm ID column at its optimum flow rate.