Capillary columns, Rtx®-BAC1 (fused silica)
Supplier: Restek
Certificates
About this item
Application-specific columns for blood alcohol analysis - achieve baseline resolution in less than three minutes.
- Ideal for abused inhalant anaesthetics, γ-hydroxybutyrate (GHB)/γ-butyrolactone (GBL), glycols, and common industrial solvents
- Rtx®-BAC2 confirmation column provides four elution order changes under the same conditions
- Stable to 260 °C
Frequently Bought Together