Guard/retention gap columns, base-deactivated (fused silica)
Supplier: Restek
Certificates
About this item
Recommended for use with Rtx®-5 Amine, Rtx®-35 Amine, Rtx®-Volatile Amine, and Stabilwax®-DB capillary columns.
- Tested with a basic amine test mix
- Excellent inertness for basic compounds
- Batch test chromatogram included
Maximum temperature: 315 °C