Guard/retention gap columns, polar-deactivated (fused silica)
Supplier: Restek
Certificates
About this item
Polyethylene glycol deactivation layer provides optimum wettability for polar compounds.
- Tested with a comprehensive test mix to ensure high inertness
- Minimize peak splitting when using polar solvents such as methanol or water
- Compatible with Stabilwax, Rtx-225, and Rt-2330 capillary columns
Maximum temperature: 280 °C