Graphite sealing rings
: Restek
These graphite sealing rings are suitable for Thermo Scientific TRACE, 8000, 8000 TOP and Focus SSL instruments. They fit liners with 8 mm outer diameter.
These graphite sealing rings are suitable for Thermo Scientific TRACE, 8000, 8000 TOP and Focus SSL instruments. They fit liners with 8 mm outer diameter.