Spill response cart, PIG® Oil-Only
Catalog # NEWPKIT404-01
Supplier: NEW PIG
Specifications
- Description:Spill response cart, PIG® Oil-Only
- Pk:1
Specifications
About this item
These large, fully-equipped kits control and absorb high-volume spills. The PE lockable container keeps absorbents clean, dry and secure before use.
- Tamperproof seals help prevent pilfering of spill response supplies
- Four-way forklift entry for easy moving
- Prepacked, compartmentalised container provides easy access to contents
- Temporary disposal bags help make cleanup easier
For use with oil based liquids and not suitable for water.
Certifications: Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables.
Delivery: Oil-Only booms of 2× (200 mm×3 m), 3× (130 mm×3 m) and 15× (80 mm×3 m), STAT-MAT® heavy-weight pads, 6× sump skimmer pillows (200×460 mm), 2× spaghetti strips, 30× disposal bags (910×1520 mm), instruction manual, 6× tamperproof labels (55×150 mm), PE box with lid (1120×1220×775 mm ) and 4× rubber wheels (Ø 13 mm).