Direct injection liners, Uniliner®
Supplier: Restek
Certificates
About this item
Direct injection Uniliner® liners minimise solvent and peak tailing. These are suitable for 6,35 mm packed column injection ports.
- Press-Tight® taper forms dead volume free connection to columns
- Can be used with 0,25, 0,32 and 0,53 mm ID capillary columns
- Can be removed from the septum nut weldment