ContactUsLinkComponent
LEADING LIGHT® Wnt Reporter Assay Starter Kit
LEADING LIGHT® Wnt Reporter Assay Starter Kit
Catalog # ENZOENZ610010001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
LEADING LIGHT® Wnt Reporter Assay Starter Kit
Catalog # ENZOENZ610010001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    LEADING LIGHT® Wnt reporter assay starter kit
  • Seq:
    02
Specifications
About this item
The LEADING LIGHT® Wnt Reporter Assay Starter Kit is a cell-based luciferase activity test suitable for 96 well plate format.
  • No transfection required - cell line, media, reagents and controls all included
  • True end-point detection system - intact pathway, allowing investigation of the complete pathway
  • High sensitivity (Wnt3a EC₅₀=45,9 ng/ml) and excellent reproducibility (Z'-factor of 0,74)
  • High signal:noise ratio without the need for lithium chloride to boost the signal
  • High throughput screening applications
The system contains an engineered 3T3 mouse fibroblast cell line, which expresses the firefly luciferase reporter gene under the control of Wnt-responsive promoters (TCF/LEF). The luciferase activity from the reporter gene in this cell line can be up-regulated in a dose-dependent manner upon the addition of exogenous Wnt protein/Wnt agonist or down-regulated by a further addition of a Wnt antagonist to the cell culture medium. This system can be used to elucidate the functions/activities of different Wnt-related ligands such as Wnt, DKK, etc. This system can also be used for screening small molecules and antibodies for their ability to act as Wnt inhibitors or Wnt agonists.
The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway.
Delivery: The kit contains LEADING LIGHT® Wnt reporter cell line, cell assay and growth medium concentrate, cell freezing medium, luciferase substrate and assay buffer, HEPES, Wnt3a protein (positive control), and Dkk-1 protein.