ContactUsLinkComponent
Scalpel blade remover, Feather®
Scalpel blade remover, Feather®
Catalog # FEAT201090000
Supplier:  PFM Medical GmbH
Scalpel blade remover, Feather®
Catalog # FEAT201090000
Supplier:  PFM Medical GmbH
Specifications
  • Description:
    Scalpel blade remover, disposable, standard size, blue
  • Pk:
    1
Specifications
About this item
This disposable blade remover permits the safe and simple removal of used disposable blades from any kind and size of scalpel handle. It makes blade changes easy and ensures increased safety and hygiene because direct contact with the blade is avoided.
  • Easy handling in three steps
  • No direct contact with blades provides the greatest possible protection of medical staff against infection
  • Once the blade remover is filled with used blades, it can simply be disposed