Dissecting blade handle and blades, FEATHER®
Supplier: PFM Medical GmbH
Certificates
About this item
The functional handle is designed for easy blade replacement and cleaning. Extra-thick stainless steel blade provides high rigidity and extra long life.
- Special heavy-duty blades, individually packed and sterile
- Two versions of blades
- Simple blade change and easy to clean
- Autoclavable