ContactUsLinkComponent
Microtome blade holders, FEATHER®
Microtome blade holders, FEATHER®
Catalog # FEAT207000160
Supplier:  PFM Medical GmbH
undefined
Microtome blade holders, FEATHER®
Catalog # FEAT207000160
Supplier:  PFM Medical GmbH
Specifications
  • Description:
    Microtome blade holder, 160
  • Length:
    160 mm
  • Material:
    Stainless steel
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
One-touch microtome blade holders for use with disposable microtome blades for paraffin and cryostat sectioning in a wide range of individual applications in routine practice, research and industry.
  • Blades can be easily replaced without tightening any screws
  • Blade insertion and replacement are equally fast and simple