Order Entry
Germany
ContactUsLinkComponent
Absorbent granules, PIG® PEAT
Absorbent granules, PIG® PEAT
Catalog # NEWPPLP404
Supplier:  NEW PIG
undefined
Absorbent granules, PIG® PEAT
Catalog # NEWPPLP404
Supplier:  NEW PIG

Specifications

  • Description:
    Absorbent granules PIG® PEAT, brown, 4,99 kg
  • Absorbency:
    30 l/bag
  • Packed:
    1 each
  • Pk:
    1

Specifications

About this item

All-natural peat moss absorbent pulls in oil-based liquids but not water. The super-absorbent, non-leaching activated sphagnum peat moss absorbs up to 30-litres of oil per bag. It is made for in and outdoor use and even removes oily sheens on floors.

  • Incinerable to help reduce waste
  • Non-abrasive composition will not harm floors or expensive machinery
  • Silica-free composition does not pose the health hazards associated with breathing in clay dust
  • Lightweight absorbent is easier than clay to carry and use without injury
Caution: Not recommended for oil spills on water or for use with corrosive liquids.