Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid ≥98%, Ultrapure
Catalog # BMOLFA012-0010
Supplier: BIOMOL RESEARCH LABORATORIES
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Molecular Formula:C22H34O2
- Molecular Weight:330.5 g/mol
- Source:Synthetic
- CAS:24880-45-3
- Purity:≥ 98%
- Appearance:Clear to pale yellow oil
- Solubility:Soluble in ethanol, hexane and methylene chloride
Specification Test Results
About this item
:
:330.512 g/mol
CAS Number:24880-45-3
Formula:C₂₂H₃₄O₂