Capillary columns, Rtx®-Dioxin2 (fused silica)
Supplier: Restek
Certificates
About this item
These capillary columns are isomer specificity for 2,3,7,8-TCDD and 2,3,7,8-TCDF achieved with one GC column. Unique selectivity for toxic dioxin and furan congeners which allows to use as a confirmation GC column.
- Thermally stable to 340 °C for longer lifetime
Specifications
- Catalog No:
- REST10759
- REST10758
- Temperature limit:
- +20...+320/340 °C
- +20...+320/340 °C