Rat recombinant biliverdin reductase
Catalog # STSSOSP-400B
Supplier: Enzo Life Sciences (Stressgen)
CAS Number:
Specifications
- Pk:20 µG
- Enzyme type:Recombinant
- Species:Rat
- Enzyme name:Biliverdin Reductase
- Enzyme synonyms:BLVR
Specifications
About this item
Biliverdin reductase (phosphoprotein BVR) is an oxireductase which catalyses the reduction of the gamma-meso bridge of biliverdin to bilirubin.