PIG® Essentials Pro chemical mat pads
Catalog # 121-0192
Supplier: NEW PIG
Specifications
- Description:Chemical mat pads, heavyweight
- L×W:480×380 mm
- Seq:01
- Absorbency:100 L
- Packed:100 pads/bag
- Pk:100
Specifications
About this item
Chemical Mat that quickly absorbs acids, caustics and unknown liquids for effective clean-up of high volume spills.
- 100% polypropylene mat quickly absorbs nearly any type of caustic, hazardous or unknown liquid- including acids, bases, solvents and chemicals
- Colour-coded safety yellow for fast identification; know exactly what you're reaching for in an emergency spill situation
- Top and bottom layers of fine fibre are built of densely woven, small diameter strands of fibre to increase surface area-enabling the mat to quickly soak up liquids on contact before they can spread
- Random assembly of large-diameter melt-blown polypropylene fibres creates a high loft inner layer that absorbs high volumes of liquid fast
- Heavy weight construction offers the greatest sorbency
Pads are ideal for catching drips and soaking up spills. Easy tear perforations let you take only what you need so you use less mat and save money.