ContactUsLinkComponent
Column kit, Stx-CLPesticides
Column kit, Stx-CLPesticides
  REST11190
 :  Restek
undefined
Column kit, Stx-CLPesticides
  REST11190
 :  Restek
  • Column Format:
    Capillary
  • Description:
    Stx-CLPesticides column kit, 0,25 mm I-Ø
  • Pk:
    1
Application specific columns for organochlorine pesticides and herbicides.
  • Baseline separations in less than 10 minutes
  • Temperature range: stable to 330 °C
  • Siltek surface deactivation enhances responses for endrin, DDT, methoxychlor
Siltek passivation technology enables these columns to offer unsurpassed inertness and the highest responses for active pesticides.
 : Kit contains an Stx-CLPesticides column, Stx-CLPesticides2 column, Universal angled 'Y' press-tight connector, and a Siltek-deactivated guard column. Columns are not preconnected in this kit.