Orders LinkContactUsLinkComponent
<i>Clostridium difficile</i> toxin B
Clostridium difficile toxin B
Clostridium difficile toxin B, 50 µg, 50 µG
Catalog # ENZOBMLG1500050
Supplier:   Enzo Life Sciences
<i>Clostridium difficile</i> toxin B
Catalog # ENZOBMLG1500050
Supplier:  Enzo Life Sciences
CAS Number:  
Clostridium difficile toxin B
Clostridium difficile toxin B, 50 µg, 50 µG
CAS Number:  
Price
1 210,00 €
Log in for availability
1 * 50 µG
Product Details & Documents
Clostridium difficile toxin B from Clostridium difficile, strain VPI10463 is a high purity toxin applicable to ELISA, antibody capture, cytotoxicity studies and LINE assay.
  • High potency retained following inactivation
  • Minimal lot-to-lot variation
  • Lyophilised for ease of handling and long term stability
Specifications
Specifications
  • Pk:
    50 µG
  • Description:
    Clostridium difficile toxin B, 50 µg