Capillary columns, Rxi®-20 (fused Silica)
Supplier: Restek
Certificates
About this item
Rtx®-20 columns are general purpose columns for volatile compounds, flavour compounds, alcoholic beverages.
- Low to midpolarity phase; Crossbond® diphenyl dimethyl polysiloxane
- Temperature stability up to 320 °C
- Equivalent to USP G28, G32 phases
Rtx®-20 polymer is synthesised to exacting standards. All residual catalysts and low molecular weight fragments are removed from the polymer, providing a tight monomodal distribution and low bleed.