Anti-IgG Goat Antibody
Catalog # JACK115-225-044
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Host:Goat
- Reactivity:Mouse
- Pk:2 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse