Anti-IgG Goat Antibody (Cy3®)
Catalog # JACK115-165-072
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Cy3®
- Host:Goat
- Reactivity:Mouse
- Pk:1,5 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy3®
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse