Anti-IgG Goat Antibody
JACK115-075-068
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Host:Goat
- Reactivity:Mouse
- Pk:1,5 mg
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse