Anti-IgG Rabbit Antibody (Cy3®)
JACK315-165-003
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Cy3®
- Host:Rabbit
- Reactivity:Mouse
- Pk:1,5 mg
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse