Anti-IgG Rabbit Antibody (Cy2®)
JACK315-225-048
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Cy2®
- Host:Rabbit
- Reactivity:Mouse
- Pk:1 mg
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy2®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse