Anti-IgM Rabbit Antibody (Cy5®)
Catalog # JACK315-175-020
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin M
- Antigen symbol:IgM
- Conjugation:Cy5®
- Host:Rabbit
- Reactivity:Mouse
- Pk:1,5 mg
Specifications
About this item
Type:
Antigen: IgM
Clonality:
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse