Anti-IgG Rat Antibody (Texas Red®)
JACK415-075-166
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Texas Red®
- Host:Rat
- Reactivity:Mouse
- Pk:1 mg
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse