Anti-IgG Sheep Antibody
Catalog # JACK515-005-003
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Host:Sheep
- Reactivity:Mouse
- Epitope:Heavy and Light Chain
- Pk:2 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope: Heavy and Light Chain
Host: Sheep
Isotype:
Reactivity: Mouse