Anti-IgG Rat Antibody (Cy2®)
Catalog # JACK415-225-166
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Cy2®
- Host:Rat
- Reactivity:Mouse
- Pk:1 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy2®
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse