Anti-IgG Rat Antibody
JACK415-155-166
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Host:Rat
- Reactivity:Mouse
- Pk:1 mg
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse