Anti-Ig Sheep Antibody (Texas Red®)
Catalog # JACK515-075-071
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin
- Antigen symbol:Ig
- Conjugation:Texas Red®
- Host:Sheep
- Reactivity:Mouse
- Pk:1,5 mg
Specifications
About this item
Type:
Antigen: Ig
Clonality:
Clone:
Conjugation: Texas Red®
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse