Anti-IgG Sheep Antibody
JACK515-175-071
Marketsource
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse
- Cat. No.:JACK515-175-071
- Reactivity:Mouse
- Host:Sheep
- Antigen symbol:IgG
- Antigen name:Immunoglobulin G
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



