Anti-IgG Donkey Antibody
Catalog # JACK715-065-140
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Host:Donkey
- Reactivity:Mouse
- Pk:0,5 mL
Specifications
About this item
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Donkey
Isotype:
Reactivity: Mouse