Anti-IgG Donkey Antibody (Cy3®)
JACK715-165-150
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Cy3®
- Host:Donkey
- Reactivity:Mouse
- Pk:0,5 mg
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy3®
Epitope:
Host: Donkey
Isotype:
Reactivity: Mouse