Anti-IgM Goat Antibody
Catalog # JACK115-175-0750
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin M
- Antigen symbol:IgM
- Host:Goat
- Reactivity:Mouse
- Pk:1,5 mg
Specifications
About this item
Type:
Antigen: IgM
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse