Anti-WDSOF1 Rabbit Polyclonal Antibody
NOVUNBP1-70747
- Antibody type:Primary
- Antigen name:DDB1 and CUL4 associated Factor 13
- Antigen symbol:WDSOF1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:50 μg
- Pk:50 µG
Rabbit Polyclonal WDSOF1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: WDSOF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human