Anti-Dot1L Rabbit Polyclonal Antibody
Catalog # NOVUNB100-40845
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:DOT1-Like
- Antigen symbol:DOT1L
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Rabbit Polyclonal Dot1L Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.
Type: Primary
Antigen: Dot1L
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human