Anti-CDH11 Mouse Monoclonal Antibody
NOVUNBP1-97676
- Antibody type:Primary
- Antigen name:Cadherin 11
- Antigen symbol:CDH11
- Clonality:Monoclonal
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:OB|Cadherin 11|OSF-4|OB-Cadherin (osteoblast)|OB-Cadherin|Cadherin-11|CDHOB|CAD11|osteoblast Cadherin|type 2
- Size:100 μg
- Pk:100 µG
Mouse Monoclonal Cadherin-11 Antibody (16A). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen. Tested Reactivity: Human, Rat.
Type: Primary
Antigen: CDH11
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: