Anti-GYS1 Rabbit Polyclonal Antibody
Catalog # NOVUNB600-611
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Glycogen Synthase 1
- Antigen symbol:GYS1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:Glycogen synthase 1 phospho Ser645|EC 2.4.1.11|UDP glucose glycogen glucosyltransferase|GYS 1|GYS1_HUMAN|p-Glycogen synthase 1 S645|Glycogen synthase1|GSY|Starchsynthase muscle|GYS|Glycogen synthase 1|Glycogen [starch] synthase|GYS1|muscl.|Glycogen synthase 1 phospho S645|Glycogen synthase 1 muscle
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Rabbit Polyclonal Glycogen Synthase [p Ser640] Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry. Tested Reactivity: Human, Mouse.
Type: Primary
Antigen: Glycogen Synthase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human