About this item
Description: The RM134L monoclonal antibody reacts with mouse CD252 also known as OX-40 Ligand, a member of the TNF superfamily. OX-40L is induced on mouse splenic B cells stimulated with a combination of anti-IgM and anti-CD40. Neither resting nor activated mouse T cells express OX-40L. The interaction of OX-40 (CD134) with OX-40L has been implicated in T cell-dependent humoral response. RM134L inhibits the costimulatory activity of mouse OX-40L transfected cells for anti-CD3-stimulated T cell proliferation. Applications Reported: This RM134L antibody has been reported for use in flow cytometric analysis. Applications Tested: This RM134L antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser The tumor necrosis factor superfamily member TNFSF4 is a type II membrane bound, non-covalently linked homotrimeric protein. It is expressed on antigen presenting cells, such as dendritic cells and activated B-cells, and also on other cells such as vascular endothelial cells, mast cells, and natural killer cells. TNFSF4 signals specifically through the TNFRSF4 receptor, is expressed predominantly on CD4+T cells but also on certain activated CD8+T cells. TNFRSF4/TNFSF4 functions as a costimulatory signal, which is required for a productive interaction between antigen presenting cells and their target T-cells. It enhances cell proliferation and survival, and increases expression of RANTES, IL-2, IL-3, and IFNgamma. TNFRSF4/TNFSF4 signaling plays an important role in immunotolerance.
Flow Cytometry: 0.5 µg/test
Type: Primary
Antigen: OX40L
Clonality: Monoclonal
Clone: RM134L
Conjugation: AP (Alkaline Phosphatase)C
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Specifications
- Catalog No:
- EBIO17-5905-80
- EBIO17-5905-82
- Antigen symbol:
- OX40L
- OX40L
- Antigen name:
- OX40 ligand
- OX40 ligand
- Antigen synonyms:
- TNFSF4|gp34|OX40 Ligand|TXGP1|Txgp1l|Ath1|CD252|Ath-1
- TNFSF4|gp34|OX40 Ligand|TXGP1|Txgp1l|Ath1|CD252|Ath-1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- RM134L
- RM134L
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 22164
- 22164
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- 0.2 mg/mL
- 0.2 mg/mL
- Storage buffer:
- PBS, 0.1% gelatin, 0.09% sodium azide, pH 7.2
- PBS, 0.1% gelatin, 0.09% sodium azide, pH 7.2
- Storage temperature:
- 4° C, store in dark, DO NOT FREEZE!
- 4° C, store in dark, DO NOT FREEZE!