Anti-CDC2 Mouse Monoclonal Antibody (Biotin) [clone: CDK1/873]
Supplier: Biotium
Certificates
About this item
Recognizes a 34 kDa protein, which is identified as cyclin dependent kinase 1 (cdk1) or p34cdc2 protein kinase. cdk1 / p34cdc2 plays a crucial role during cell division and is most active during mitosis. It is predominantly localized in the nucleus. It is a serine/threonine kinase, which is activated by cyclin, presumably by de-phosphorylation of tyrosine residues. Activated cdk1 / p34cdc2 performs specific functions during mitosis, including nuclear envelope breakdown and chromosome condensation.
Type: Primary
Antigen: CDC2
Clonality: Monoclonal
Clone: CDK1/873
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Cow, Human, Mink, Monkey
Specifications
- Catalog No:
- BNCB0873-500
- BNCB0873-100
- Antigen symbol:
- CDC2
- CDC2
- Antigen name:
- p34 / cdk1
- p34 / cdk1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone CDK1/873
- Clone CDK1/873
- Reactivity:
- Monkey, Human, Bovine, Mink
- Monkey, Human, Bovine, Mink
- Host:
- Mouse
- Mouse
- Gene ID:
- 334562
- 334562
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide