Order Entry
Germany
ContactUsLinkComponent
 
Anti-CDC2 Mouse Monoclonal Antibody (CF647) [clone: POH-1]
 :  Biotium
 
Anti-CDC2 Mouse Monoclonal Antibody (CF647) [clone: POH-1]
 :  Biotium

 

  • Catalog No:
  • BNC470853-500
  • BNC470853-100
  • Antigen symbol:
  • CDC2
  • CDC2
  • Antigen name:
  • p34 / cdk1
  • p34 / cdk1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone POH-1
  • Clone POH-1
  • Reactivity:
  • Monkey, Human, Bovine, Mink
  • Monkey, Human, Bovine, Mink
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 334562
  • 334562
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide