Order Entry
Germany
ContactUsLinkComponent
Anti-CHGA Mouse Monoclonal Antibody (Biotin) [clone: CHGA/798]
Anti-CHGA Mouse Monoclonal Antibody (Biotin) [clone: CHGA/798]
Supplier:  Biotium
Certificates
Anti-CHGA Mouse Monoclonal Antibody (Biotin) [clone: CHGA/798]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0798-100
  • BNCB0798-500
  • Antigen symbol:
  • CHGA
  • CHGA
  • Antigen name:
  • Chromogranin A
  • Chromogranin A
  • Antigen synonyms:
  • Pituitary Secretory Protein I|Chromogranin A Parathyroid Secretory Protein 1|ER-37|Vasostatin I or II|Parastatin|SP-I|Pancreastatin|CHGA|Beta-Granin|CGA
  • Pituitary Secretory Protein I|Chromogranin A Parathyroid Secretory Protein 1|ER-37|Vasostatin I or II|Parastatin|SP-I|Pancreastatin|CHGA|Beta-Granin|CGA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone CHGA/798
  • Clone CHGA/798
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 150793
  • 150793
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications