Anti-GAPex5/GAPVD1/RAP6 Rabbit Polyclonal Antibody
BOSSBS-13283R
: Bioss
- Antibody type:Primary
- Antigen symbol:GAPex5/GAPVD1/RAP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:26130
- Antigen synonyms:GTPase activating protein and VPS9 domains 1|GAPD1_HUMAN.|Rab5 activating protein 6|KIAA1521|GAPEX 5|RAP6|GAPVD1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Type: Primary
Antigen: GAPex5/GAPVD1/RAP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat