Anti-PBK Rabbit Polyclonal Antibody
Catalog # BOSSBS-3523R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:PDZ binding kinase
- Antigen symbol:PBK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:55872
- Antigen synonyms:TOPK|T-lymphokine-activated killer cell-originated protein kinase|MAPKK-like protein kinase|PBK|Nori-3|SPK|Spermatogenesis-related protein kinase|PDZ-binding kinase|T-LAK cell-originated protein kinase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:280-320/322
- Purification:Purified by Protein A
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PBK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat