Order Entry
Germany
ContactUsLinkComponent
Anti-alpha GalCer:CD1d Complex Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: L363]
Anti-alpha GalCer:CD1d Complex Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: L363]
Supplier:  E-Bioscience
Certificates
Anti-alpha GalCer:CD1d Complex Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: L363]
Supplier:  E-Bioscience

Specifications

  • Catalog No:
  • EBIO12-2019-80
  • BOSSBS-3264R
  • Antigen symbol:
  • alpha GalCer:CD1d Complex
  • ITGB2
  • Antigen name:
  • alpha GalCer:CD1d Complex
  • Integrin Beta 2
  • Antigen synonyms:
  • LFA-1|CD18|MAC-1|integrin subunit beta 2|LAD|LCAMB|MFI7|MF17
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Polyclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • Unconjugated
  • Clone:
  • L363
  • Reactivity:
  • Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Rabbit
  • Gene ID:
  • 12479
  • 3689
  • Isotype:
  • IgG2a kappa
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • Flow cytometry:
  • Yes
  • Modification:
  • phospho-Ser756/Thr758/Thr759
  • Immunogen:
  • 724-769/769
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Affinity chromatography
  • Purified by Protein A
  • Concentration:
  • 0.2 mg/mL
  • 1 ug/ul
  • Cross adsorption:
  • No
  • Storage buffer:
  • PBS, 0.1% gelatin, 0.09% sodium azide, pH 7.2
  • Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Storage temperature:
  • 4° C, store in dark, DO NOT FREEZE!
  • Store at –20 °C for 12 months
  • Shipping temperature:
  • 4 °C

Specifications

Related Information