Restriction enzymes, BsmBI, FastDigest™, fermentas
Catalog # FD0454
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Compatible buffer:FastDigest Green Buffer or FastDigest Buffer (colorless)
- Overhang type:5' overhang
- Description:BsmBI, FastDigest™
- Enzyme name:FastDigest Esp3I
- No. of reactions:20 (20 µl)
Specifications
About this item
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Designed to complete digestion in 5 to 15 minutes
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications